missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCRL2/LCCR Antibody (498321), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB5519CL1
This item is not returnable.
View return policy
Description
CCRL2/LCCR Monoclonal antibody specifically detects CCRL2/LCCR in Mouse samples. It is validated for Flow Cytometry,Immunocytochemistry,CyTOF
Specifications
| CCRL2/LCCR | |
| Monoclonal | |
| CoraFluor 1 | |
| ACKR5;C-C chemokine receptor-like 2;CKRX;CRAM;CRAM-A;CRAM-B;HCR | |
| RBL-1 rat basophilic leukemia cell line transfected with mouse CCRL2/LCCR, Accession # O35457 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 498321 | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 9034 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction