missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCRL2/CRAM-A/B Rabbit anti-Mouse, Clone: BZ5B8, Novus Biologicals™
Rabbit Monoclonal Antibody
£210.00 - £463.00
Specifications
| Antigen | CCRL2/CRAM-A/B |
|---|---|
| Clone | BZ5B8 |
| Dilution | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18364372
|
Novus Biologicals
NBP3-11970-0.025MG |
0.025 mg |
£210.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18329501
|
Novus Biologicals
NBP3-11970-0.2MG |
0.2 mg |
£463.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCRL2/CRAM-A/B Monoclonal antibody specifically detects CCRL2/CRAM-A/B in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Functional AssaySpecifications
| CCRL2/CRAM-A/B | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Functional Assay | |
| Unconjugated | |
| RUO | |
| Mouse | |
| C-C chemokine receptor-like 2, CCR11, CCR6, chemokine (C-C motif) receptor-like 2, Chemokine receptor CCR11, Chemokine receptor X, CKRXMGC116710, CRAM, CRAM-A, CRAM-B, FLJ55815, HCRMGC34104, Putative MCP-1 chemokine receptor | |
| This antibody was raised by immunising Wistar Furth rats with the mCCRL2/CRAM-A/B peptide/KLH conjugate. The immunizing amino-terminal mCCRL2/CRAM-A/B peptide with the sequence NH2-MDNYTVAPDDEYDVLILDDYLDNSC-COOH, corresponding to residues 1-24 of mCCRL2/CRAM-A/B, with a nonnative carboxyl-terminal cysteine to facilitate conjugation to keyhole limpet hemocyanin (KLH). | |
| Primary |
| BZ5B8 | |
| Monoclonal | |
| Rabbit | |
| GPCR, Immunology, Innate Immunity | |
| PBS | |
| 9034 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title