missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCRL2/CRAM-A/B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15769
This item is not returnable.
View return policy
Description
CCRL2/CRAM-A/B Polyclonal specifically detects CCRL2/CRAM-A/B in Human, Mouse samples. It is validated for Western Blot.
Specifications
| CCRL2/CRAM-A/B | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| C-C chemokine receptor-like 2, CCR11, CCR6, chemokine (C-C motif) receptor-like 2, Chemokine receptor CCR11, Chemokine receptor X, CKRXMGC116710, CRAM, CRAM-A, CRAM-B, FLJ55815, HCRMGC34104, Putative MCP-1 chemokine receptor | |
| Rabbit | |
| 40 kDa | |
| 0.1 mL | |
| GPCR, Immunology, Innate Immunity | |
| 9034 | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| 0.29mg/mL | |
| Western Blot 1:500-1:3000 | |
| O00421 | |
| CCRL2 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human CCRL2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse. Expected to cross react based on sequence identity: Human (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction