missing translation for 'onlineSavingsMsg'
Learn More

CCRL2/CRAM-A/B Antibody (BZ2E3), DyLight 594, Novus Biologicals™

Product Code. 18370842 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370842 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18370842 Supplier Novus Biologicals Supplier No. NBP311982DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CCRL2/CRAM-A/B Monoclonal antibody specifically detects CCRL2/CRAM-A/B in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCRL2/CRAM-A/B
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone BZ2E3
Conjugate DyLight 594
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias C-C chemokine receptor-like 2, CCR11, CCR6, chemokine (C-C motif) receptor-like 2, Chemokine receptor CCR11, Chemokine receptor X, CKRXMGC116710, CRAM, CRAM-A, CRAM-B, FLJ55815, HCRMGC34104, Putative MCP-1 chemokine receptor
Host Species Rabbit
Immunogen This antibody was raised by immunising Wistar Furth rats with the mCCRL2/CRAM-A/B peptide/KLH conjugate. The immunizing amino-terminal mCCRL2/CRAM-A/B peptide with the sequence NH2-MDNYTVAPDDEYDVLILDDYLDNSC-COOH, corresponding to residues 1-24 of mCCRL2/CRAM-A/B, with a nonnative carboxyl-terminal cysteine to facilitate conjugation to keyhole limpet hemocyanin (KLH).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 9034
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.