missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCRL2/CRAM-A/B Antibody (152211), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2350C
This item is not returnable.
View return policy
Description
CCRL2/CRAM-A/B Monoclonal antibody specifically detects CCRL2/CRAM-A/B in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| CCRL2/CRAM-A/B | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 152211 | |
| Flow Cytometry, CyTOF-ready | |
| C-C chemokine receptor-like 2, CCR11, CCR6, chemokine (C-C motif) receptor-like 2, Chemokine receptor CCR11, Chemokine receptor X, CKRXMGC116710, CRAM, CRAM-A, CRAM-B, FLJ55815, HCRMGC34104, Putative MCP-1 chemokine receptor | |
| NS0 mouse myeloma cell line transfected with human CCRL2/CRAM-A/B, Met1-Val356 (Val180Met), Accession # NP_001124382 | |
| 0.5 mL | |
| GPCR, Immunology, Innate Immunity | |
| 9034 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction