missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR9 Rabbit anti-Mouse, Clone: 9B1, Novus Biologicals™
Rabbit Monoclonal Antibody
£191.00 - £510.00
Specifications
| Antigen | CCR9 |
|---|---|
| Clone | 9B1 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry, Immunocytochemistry/Immunofluorescence, Neutralization |
| Applications | Flow Cytometry, Immunofluorescence, Neutralization |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18303232
|
Novus Biologicals
NBP3-12001-0.025MG |
0.025 mg |
£191.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18338181
|
Novus Biologicals
NBP3-12001-0.2MG |
0.2 mg |
£510.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR9 Monoclonal antibody specifically detects CCR9 in Mouse samples. It is validated for Flow Cytometry, Immunofluorescence, NeutralizationSpecifications
| CCR9 | |
| 1 mg/mL | |
| Flow Cytometry, Immunofluorescence, Neutralization | |
| Unconjugated | |
| RUO | |
| Mouse | |
| C-C chemokine receptor type 9, C-C CKR-9, CC-CKR-9, CCR-9, CDw199, CDw199 antigen, chemokine (C-C motif) receptor 9, G protein-coupled receptor 28, GPR28, GPR-9-6G-protein coupled receptor 28 | |
| This antibody was raised by immunizing rats with KLH-conjugated linear peptide corresponding to the N-terminus of mouse CCR9. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| 9B1 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence, Neutralization | |
| Monoclonal | |
| Rabbit | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| PBS | |
| 10803 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title