missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR9 Antibody (882416) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8287C
This item is not returnable.
View return policy
Description
CCR9 Monoclonal antibody specifically detects CCR9 in Rat samples. It is validated for Flow Cytometry, CyTOF
Specifications
| CCR9 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
| Flow Cytometry, CyTOF | |
| 882416 | |
| Flow Cytometry, CyTOF-ready | |
| C-C chemokine receptor type 9, C-C CKR-9, CC-CKR-9, CCR-9, CDw199, CDw199 antigen, chemokine (C-C motif) receptor 9, G protein-coupled receptor 28, GPR28, GPR-9-6G-protein coupled receptor 28 | |
| NS0 mouse myeloma cell line transfected with rat CCR9 | |
| 0.1 mL | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 10803 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction