missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CCR8 Polyclonal Antibody
GREENER_CHOICE

Sheep Polyclonal Antibody

Brand:  Invitrogen™ PA5149109

Product Code. 30535702

  • £407.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

CCR8 encodes a member of the beta chemokine receptor family, which is predicted to be a seven transmembrane protein similar to G protein-coupled receptors. Chemokines and their receptors are important for the migration of various cell types into the inflammatory sites. This receptor protein preferentially expresses in the thymus. I-309, thymus activation-regulated cytokine (TARC) and macrophage inflammatory protein-1 beta (MIP1 beta) have been identified as ligands of this receptor. Studies of this receptor and its ligands suggested its role in regulation of monocyte chemotaxis and thymic cell apoptosis. More specifically, this receptor may contribute to the proper positioning of activated T cells within the antigenic challenge sites and specialized areas of lymphoid tissues. This gene is located at the chemokine receptor gene cluster region. CCR8 expression has been reported in spleen and thymus, natural-killer cells, monocytes, and T cells, and at lower levels in peripheral blood leukocytes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CCR8
Polyclonal
Unconjugated
CCR8
C-C; CC chemokine receptor 8; CC chemokine receptor CHEMR1; C-C chemokine receptor type 8; C-C CKR-8; C-C motif chemokine receptor 8; CC-chemokine receptor chemr1; CC-CKR-8; CCR8; CCR-8; cd198; CDw198; chemokine (C-C motif) receptor 8; chemokine (C-C) receptor 8; chemokine (C-C) receptor-like 2; chemokine receptor-like 1; CKR-8; CKRL1; CKR-L1; Cmkbr8; CMKBRL2; CY6; GPRCY6; GPR-CY6; mCCR8; MGC129966; MGC129973; TER1
Sheep
Antigen affinity chromatography
RUO
12776, 301066
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Western Blot
0.2 mg/mL
PBS with trehalose and no preservative
P56484
CCR8
E. coli-derived recombinant mouse CCR8, Met1-Met33, Asn92-Lys105, Tyr170-Ile200, His262-Leu278.
100 μg
Primary
Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.