missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR8 (CD198) Mouse anti-Human , BB515, Clone: 433H, BD Biosciences™
Mouse Monoclonal Antibody
£187.65 - £299.60
Specifications
| Antigen | CCR8 (CD198) |
|---|---|
| Clone | 433H |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Blue 515 (BB515) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17287684
|
BD Biosciences
566892 |
25 Tests |
£196.00 £187.65 / 25 tests Save £8.35 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17277684
|
BD Biosciences
566891 |
100 Tests |
£312.00 £299.60 / 100 tests Save £12.40 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR8 (CD198) Monoclonal antibody specifically detects CCR8 (CD198) in Human samples. It is validated for Flow CytometrySpecifications
| CCR8 (CD198) | |
| Flow Cytometry | |
| Brilliant Blue 515 (BB515) | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human CCR8 Transfected Cell Line | |
| Primary |
| 433H | |
| Monoclonal | |
| Mouse | |
| Human | |
| CCR8; CKR-L1; CKRL1; CMKBR8; CMKBRL2; CY6; GPR-CY6; GPRCY6; MGC129966; MGC1 | |
| IgG2a κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title