missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR7 Rabbit anti-Mouse, Clone: 4B12, Novus Biologicals™
Rabbit Monoclonal Antibody
£209.00 - £456.00
Specifications
| Antigen | CCR7 |
|---|---|
| Clone | 4B12 |
| Concentration | 1 mg/ml |
| Dilution | Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Functional Assay |
| Applications | Flow Cytometry, Immunofluorescence, Immunoprecipitation, Functional Assay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18301892
|
Novus Biologicals
NBP3-12012-0.025MG |
0.025 mg |
£209.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18043784
|
Novus Biologicals
NBP3-12012-0.2MG |
0.2 mg |
£456.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR7 Monoclonal antibody specifically detects CCR7 in Mouse samples. It is validated for Flow Cytometry, Immunofluorescence, Immunoprecipitation, Functional AssaySpecifications
| CCR7 | |
| 1 mg/ml | |
| Flow Cytometry, Immunofluorescence, Immunoprecipitation, Functional Assay | |
| Unconjugated | |
| Rabbit | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| PBS | |
| 1236 | |
| IgG κ | |
| Protein A purified |
| 4B12 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Functional Assay | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| BLR2C-C chemokine receptor type 7, C-C CKR-7, CC-CKR-7, CCR-7, CD197, CD197 antigen, CDw197CC chemokine receptor 7, chemokine (C-C motif) receptor 7, CMKBR7chemokine (C-C) receptor 7, EBI1EBV-induced G protein-coupled receptor 1, EBV-induced G-protein coupled receptor 1, Epstein-Barr virus induced gene 1, Epstein-Barr virus induced G-protein coupled receptor, Epstein-Barr virus-induced G-protein coupled receptor 1, EVI1, lymphocyte-specific G protein-coupled peptide receptor, MIP-3 beta receptor | |
| This antibody was raised by immunizing rats with mouse CCR7 antigen. | |
| Primary | |
| Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title