missing translation for 'onlineSavingsMsg'
Learn More

CCR7 (CD197) Mouse anti-Human , BV650, Clone: 2-L1-A, BD Biosciences™

Mouse Monoclonal Antibody

Brand:  BD Biosciences 566756

 View more versions of this product

Product Code. 17237584

  • ONLINE EXCLUSIVE PRICE
    £412.00
    £395.47 / 100 tests
    Save £16.53
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CCR7 (CD197) Monoclonal antibody specifically detects CCR7 (CD197) in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CCR7 (CD197)
Monoclonal
Brilliant Violet 650
BLR2; CC chemokine receptor 7; CMKBR7; EBI1; EVI1; Epstein-Barr virus induced gene 1; MIP-3 beta receptor
Human CCR7 Transfected Cell Line
RUO
Human
IgG1 κ
Flow Cytometry
2-L1-A
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
100 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV650 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV650 were removed.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.