missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR7 (CD197) Mouse anti-Human , BV510, Clone: 2-L1-A, BD Biosciences™
Mouse Monoclonal Antibody
£195.48 - £407.22
Specifications
| Antigen | CCR7 (CD197) |
|---|---|
| Clone | 2-L1-A |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Violet 510 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17287584
|
BD Biosciences
566761 |
25 Tests |
£198.00 £195.48 / 25 tests Save £2.52 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17277584
|
BD Biosciences
566760 |
100 Tests |
£412.00 £407.22 / 100 tests Save £4.78 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR7 (CD197) Monoclonal antibody specifically detects CCR7 (CD197) in Human samples. It is validated for Flow CytometrySpecifications
| CCR7 (CD197) | |
| Flow Cytometry | |
| Brilliant Violet 510 | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human CCR7 Transfected Cell Line | |
| Primary |
| 2-L1-A | |
| Monoclonal | |
| Mouse | |
| Human | |
| BLR2; CC chemokine receptor 7; CMKBR7; EBI1; EVI1; Epstein-Barr virus induced gene 1; MIP-3 beta receptor | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV510 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV510 were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title