missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
CCR7 Antibody (3D12) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
£157.00 - £377.00
Tekniske data
| Antigen | CCR7 |
|---|---|
| Clone | 3D12 |
| Concentration | 0.5 mg/mL |
| Dilution | Flow Cytometry < = 0.5 ug/10^6 cells in 100 ul, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry, Flow Cytometry |
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|
18261704
|
Novus Biologicals
NBP1-43332-25UG |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18257494
|
Novus Biologicals
NBP1-43332-0.1MG |
0.1 mg |
£377.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
CCR7 Monoclonal specifically detects CCR7 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Tekniske data
| CCR7 | |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Immunohistochemistry, Flow Cytometry | |
| Unconjugated | |
| RUO | |
| Human | |
| P32248 | |
| 1236 | |
| The immunogen for this antibody was CCR7. | |
| Primary | |
| Store at 4C. Do not freeze. |
| 3D12 | |
| Flow Cytometry < = 0.5 ug/10^6 cells in 100 ul, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Monoclonal | |
| Rat | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| PBS, pH 7.2 with 0.09% Sodium Azide | |
| BLR2C-C chemokine receptor type 7, C-C CKR-7, CC-CKR-7, CCR-7, CD197, CD197 antigen, CDw197CC chemokine receptor 7, chemokine (C-C motif) receptor 7, CMKBR7chemokine (C-C) receptor 7, EBI1EBV-induced G protein-coupled receptor 1, EBV-induced G-protein coupled receptor 1, Epstein-Barr virus induced gene 1, Epstein-Barr virus induced G-protein coupled receptor, Epstein-Barr virus-induced G-protein coupled receptor 1, EVI1, lymphocyte-specific G protein-coupled peptide receptor, MIP-3 beta receptor | |
| CCR7 | |
| IgG2a κ | |
| Affinity Purified |
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel