missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR5 Rabbit anti-Human, Clone: V14/2, Novus Biologicals™
Rabbit Monoclonal Antibody
£209.00 - £456.00
Specifications
| Antigen | CCR5 |
|---|---|
| Clone | V14/2 |
| Dilution | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18359263
|
Novus Biologicals
NBP3-11996-0.025MG |
0.025 mg |
£209.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18076825
|
Novus Biologicals
NBP3-11996-0.2MG |
0.2 mg |
£456.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR5 Monoclonal antibody specifically detects CCR5 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence, ImmunoprecipitationSpecifications
| CCR5 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Human | |
| C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5 | |
| This antibody was raised by immunizing mice with a synthetic phospho peptide corresponding to the C-Terminal region of human CCR5 conjugated to BSA. | |
| Primary |
| V14/2 | |
| Monoclonal | |
| Rabbit | |
| Alzheimers Research, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction | |
| PBS | |
| 1234 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title