missing translation for 'onlineSavingsMsg'
Learn More

CCR5, p Ser337 Antibody (V14/2), FITC, Novus Biologicals™

Product Code. 18387213 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18387213 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18387213 Supplier Novus Biologicals Supplier No. NBP311997F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCR5 Monoclonal antibody specifically detects CCR5 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCR5
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone V14/2
Conjugate FITC
Dilution Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5
Host Species Mouse
Immunogen This antibody was raised by immunizing mice with a synthetic phospho peptide corresponding to the C-Terminal region of human CCR5 conjugated to BSA.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1234
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.