missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR5 Antibody (T21/8), HRP, Novus Biologicals™
Description
CCR5 Monoclonal antibody specifically detects CCR5 in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | CCR5 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | T21/8 |
| Conjugate | HRP |
| Dilution | Western Blot, ELISA |
| Formulation | PBS |
| Gene Alias | C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5 |
| Host Species | Rabbit |
| Immunogen | This antibody was raised by immunizing mice with intact RBL-CCR5 cells. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?