missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
£187.00 - £426.00
Specifications
| Antigen | CCR5 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 2 ug/ml, Simple Western 1:50, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18755553
|
Novus Biologicals
NBP2-31374SS |
0.025 mg |
£187.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18711544
|
Novus Biologicals
NBP2-31374 |
0.1 mg |
£426.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR5 Polyclonal specifically detects CCR5 in Human, Mouse, Monkey samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CCR5 | |
| Western Blot 2 ug/ml, Simple Western 1:50, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Monkey | |
| P51681 | |
| 1234 | |
| Partial recombinant protein made to an N-terminal portion of the human CCR5 protein (between amino acids 1-350) [Uniprot: P51681] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Cancer, Chemokine Research, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction | |
| PBS with 0.05% Sodium Azide | |
| C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5 | |
| CCR5 | |
| IgG | |
| Protein A purified | |
| Immunogen has 86% homology to bovine. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title