Learn More
CCR5 Antibody (45529), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB184MFV450
400.35 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18600130
  • £468.00 / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
CCR5 Monoclonal antibody specifically detects CCR5 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CCR5
Monoclonal
mFluor Violet 450 SE
50mM Sodium Borate
Mouse
Protein A or G purified from ascites
RUO
Primary
Human
Purified
Flow Cytometry, CyTOF
45529
Flow Cytometry, CyTOF-ready
C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5
NS0 mouse myeloma cell line transfected with human CCR5, Met1-Leu352, Accession # P51681
0.1 mL
Alzheimers Research, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction
1234
Store at 4C in the dark.
IgG2b