missing translation for 'onlineSavingsMsg'
Learn More

CCR4 Antibody (KW-0761 (Mogamulizumab)) - Chimeric - Azide and BSA Free, Novus Biologicals™

Product Code. 18390450 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18390450 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18390450 Supplier Novus Biologicals Supplier No. NBP281270SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CCR4 Monoclonal specifically detects CCR4 in Human samples. It is validated for Western Blot, Functional.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCR4
Applications Western Blot, Functional Assay
Classification Monoclonal
Clone KW-0761 (Mogamulizumab)
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Functional
Formulation PBS
Gene Alias C-C chemokine receptor type 4, C-C CKR-4, CC-CKR-4HGCN:14099, CCR-4, CD194, CD194 antigen, chemokine (C-C motif) receptor 4, chemokine (C-C) receptor 4, ChemR13, CKR4MGC88293, CMKBR4k5-5, K5-5
Gene Symbols CCR4
Host Species Rabbit
Immunogen This antibody was prepared by humanization of a mouse anti-CCR4 mAb.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1233
Test Specificity This antibody is specific for human CCR4, a chemokine receptor that is preferentially expressed by Th2 and regulatory T (Treg) cells. CCR4 is also detected on basophils, platelets, monocytes and brain microvascular and coronary artery endothelial cells.
Target Species Human
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.