missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR3, Rabbit anti-Human, Clone: 5E8-G9-B4, Novus Biologicals™
Rabbit Monoclonal Antibody
£207.00 - £444.00
Specifications
| Antigen | CCR3 |
|---|---|
| Clone | 5E8-G9-B4 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18325640
|
Novus Biologicals
NBP2-81046-0.025MG |
0.025 mg |
£207.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18352150
|
Novus Biologicals
NBP2-81046-0.2MG |
0.2 mg |
£444.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR3 Monoclonal specifically detects CCR3 in Human samples. It is validated for Flow Cytometry.Specifications
| CCR3 | |
| 1 mg/mL | |
| Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| PBS | |
| 1232 | |
| Recombinant monoclonal antibody to CCR3 (CD193). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 5E8-G9-B4 | |
| Flow Cytometry | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| b-chemokine receptor, CC chemokine receptor 3, C-C chemokine receptor type 3, C-C CKR-3, CCR-3, CD193 antigen, chemokine (C-C motif) receptor 3, CKR3CC-CKR-3CMKBR3CD193, eosinophil CC chemokine receptor 3, Eosinophil eotaxin receptor, MGC102841 | |
| CCR3 | |
| IgG κ | |
| Protein A purified | |
| Recognises human CCR3, a chemokine receptor highly expressed of eosinophils, as well as Th2-Tcells, basophils, platelets, mast-cells, mononuclear phagocytes, airway epithelial cells, and hematopoietic progenitor cells (CD34+). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title