missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR3 Antibody (83103), Allophycocyanin/Cy7, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1551APCCY7
This item is not returnable.
View return policy
Description
CCR3 Monoclonal antibody specifically detects CCR3 in Mouse samples. It is validated for Flow Cytometry
Specifications
| CCR3 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 83103 | |
| Flow Cytometry | |
| b-chemokine receptor, CC chemokine receptor 3, C-C chemokine receptor type 3, C-C CKR-3, CCR-3, CD193 antigen, chemokine (C-C motif) receptor 3, CKR3CC-CKR-3CMKBR3CD193, eosinophil CC chemokine receptor 3, Eosinophil eotaxin receptor, MGC102841 | |
| Y3 rat myeloid cell line transfected with mouse CCR3, Met1-Phe359, Accession # P51678 | |
| 0.1 mL | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 1232 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction