missing translation for 'onlineSavingsMsg'
Learn More

CCR3 Antibody (5E8) - BSA Free, Novus Biologicals™

Product Code. 18382055 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18382055 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18382055 Supplier Novus Biologicals Supplier No. NBP309149

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCR3 Monoclonal specifically detects CCR3 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCR3
Applications Flow Cytometry
Classification Monoclonal
Clone 500000000
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 1-4 ug/ml
Formulation PBS
Gene Alias b-chemokine receptor, CC chemokine receptor 3, C-C chemokine receptor type 3, C-C CKR-3, CCR-3, CD193 antigen, chemokine (C-C motif) receptor 3, CKR3CC-CKR-3CMKBR3CD193, eosinophil CC chemokine receptor 3, Eosinophil eotaxin receptor, MGC102841
Host Species Mouse
Immunogen Tissue/ cell preparation (Human) - human CCR3 transfected into mouse B cell lymphoma L1.2 cells.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1232
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.