missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR2 Rabbit anti-Human, Mouse, Rat, Clone: 10L1L6, Novus Biologicals™
Rabbit Monoclonal Antibody
£167.00 - £405.00
Specifications
| Antigen | CCR2 |
|---|---|
| Clone | 10L1L6 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18307885
|
Novus Biologicals
NBP3-16117-20UL |
20 μg |
£167.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18339226
|
Novus Biologicals
NBP3-16117-100UL |
100 μg |
£405.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR2 Monoclonal antibody specifically detects CCR2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| CCR2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 729230 | |
| IgG | |
| Affinity purified |
| 10L1L6 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2CKR2B, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CCR2 (P41597). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title