missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR2 (CD192) Mouse anti-Human, Brilliant Violet 786, Clone: LS132.1D9, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 747855
This item is not returnable.
View return policy
Description
CCR2 (CD192) Monoclonal antibody specifically detects CCR2 (CD192) in Human samples. It is validated for Flow Cytometry.
Specifications
| CCR2 (CD192) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| CCR2 (CD192) | |
| Refrigerate (2 to 8°C) |
| Flow Cytometry | |
| LS132.1D9 (also known as 1D9) | |
| Brilliant Violet 786 | |
| 50 μg | |
| Primary | |
| Human | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction