missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CCR2 Antibody (plozalizumab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28546-1mg
This item is not returnable.
View return policy
Description
CCR2 Monoclonal antibody specifically detects CCR2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CCR2 | |
| Monoclonal | |
| Unconjugated | |
| P41597 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| plozalizumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2CKR2B, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor | |
| CCR2/ CD192 | |
| 1 mg | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 729230 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction