missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCR11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106552
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CCRL1.
Receptors for the C-C chemokine family include CCR-1, CCR-2A, CCR-3, CCR-4, CCR-5 and the Duffy blood group antigen. The C-C receptors are important in the function of T cell chemotaxis and migration of phagocytic cells to sites of inflammation. CCR11 in particular is expressed in the heart, lung, and small intestine. It binds members of the monocyte chemoattractant family including MCP-1, MCP-2, and MCP-4.
Specifications
| CCR11 | |
| Polyclonal | |
| Unconjugated | |
| ACKR4 | |
| A630091E18Rik; Ackr4; Atypical chemokine receptor 4; C-C chemokine receptor type 11; CC chemokine receptor-like 1; C-C CKR-11; CCBP2; CC-CKR-11; CCR10; CCR11; CCR-11; Ccrl1; CCX CKR; CCX-CKR; CCX-CKR1; chemocentryx chemokine receptor; chemokine (C-C motif) receptor-like 1; chemokine (C-C) receptor-like 1; chemokine receptor CCR11; chemokine, cc motif, receptor-like protein 1; CKR-11; Cmkbrl1; orphan seven-transmembrane receptor, chemokine related; PPR1; VSHK1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 252837, 51554, 685243 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q924I3, Q9NPB9 | |
| ACKR4 | |
| A synthesized peptide derived from human ACKR4(Accession Q9NPB9), corresponding to amino acid residues P300-I350. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction