missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR10/GPR2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NB100-56319
This item is not returnable.
View return policy
Description
CCR10/GPR2 Polyclonal specifically detects CCR10/GPR2 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Flow (Cell Surface), Flow (Intracellular).
Specifications
| CCR10/GPR2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.2% BSA with 0.05% Sodium Azide | |
| CC chemokine receptor 10, C-C chemokine receptor type 10, C-C CKR-10, CC-CKR-10, CCR-10, chemokine (C-C motif) receptor 10, G protein-coupled receptor 2, GPR2G-protein coupled receptor 2 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| In A375, a ∼50 kDa band is observed. The CCR10 antibody is made against an epitope in the cytoplasmic domain of the receptor (Moed et al, 2004). For flow cytometry (intracellular) Moed et al fixed cells with 2% formaldehye and permeabilized with 0.1% saponin. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| 0.5 mg/mL | |
| Western Blot 1-2 ug/ml, Simple Western 1:50, Flow Cytometry 1:10-1:1000, Immunohistochemistry, Immunohistochemistry-Frozen 1:10-1:2000. Use reported in scientific literature (Vestergaard et al (2003)), Flow (Cell Surface), Flow (Intracellular) reported in scientific literature (PMID 15663561) | |
| P46092 | |
| CCR10 | |
| Rabbit anti-CCR10 polyclonal antibody was raised against a peptide corresponding to amino acids 350-362 of human CCR10. In this region, mouse and human amino acid sequences are 100% identical. | |
| 0.1 mg | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 2826 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction