missing translation for 'onlineSavingsMsg'
Learn More

CCR10/GPR2 Antibody, Novus Biologicals™

Product Code. 18066954 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.10mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18066954 0.025 mg 25µg
18750754 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18066954 Supplier Novus Biologicals Supplier No. NB10056319SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 6 publications

CCR10/GPR2 Polyclonal specifically detects CCR10/GPR2 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Flow (Cell Surface), Flow (Intracellular).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCR10/GPR2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Simple Western 1:50, Flow Cytometry 1:10-1:1000, Immunohistochemistry, Immunohistochemistry-Frozen 1:10-1:2000. Use reported in scientific literature (Vestergaard et al (2003)), Flow (Cell Surface), Flow (Intracellular) reported in scientific literature (PMID 15663561)
Formulation PBS with 0.2% BSA with 0.05% Sodium Azide
Gene Accession No. P46092
Gene Alias CC chemokine receptor 10, C-C chemokine receptor type 10, C-C CKR-10, CC-CKR-10, CCR-10, chemokine (C-C motif) receptor 10, G protein-coupled receptor 2, GPR2G-protein coupled receptor 2
Gene Symbols CCR10
Host Species Rabbit
Immunogen Rabbit anti-CCR10 polyclonal antibody was raised against a peptide corresponding to amino acids 350-362 of human CCR10. In this region, mouse and human amino acid sequences are 100% identical.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2826
Test Specificity In A375, a ∼50 kDa band is observed. The CCR10 antibody is made against an epitope in the cytoplasmic domain of the receptor (Moed et al, 2004). For flow cytometry (intracellular) Moed et al fixed cells with 2% formaldehye and permeabilized with 0.1% saponin.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Vedi altri risultati Mostra meno risultati

For Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.