missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR1 Rabbit anti-Human, Clone: 3, Novus Biologicals™
Rabbit Monoclonal Antibody
£287.00 - £420.00
Specifications
| Antigen | CCR1 |
|---|---|
| Clone | 3 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:250-1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18274437
|
Novus Biologicals
NBP2-89235-50UL |
50 μg |
£287.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18249087
|
Novus Biologicals
NBP2-89235-100UL |
100 μg |
£420.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR1 Monoclonal specifically detects CCR1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CCR1 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:250-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| C-C chemokine receptor type 1, CC-CKR-1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R | |
| A synthetic peptide corresponding to the center region of the Human CCR1 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 3 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 1230 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title