missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR1 Rabbit anti-Human, Clone: 2, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89234-100UL
This item is not returnable.
View return policy
Description
CCR1 Monoclonal specifically detects CCR1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| CCR1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 2 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-4 ul/mg of lysate | |
| C-C chemokine receptor type 1, CC-CKR-1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R | |
| A synthetic peptide corresponding to the center region of the Human CCR1 | |
| 100 μg | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 1230 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction