missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR1 Antibody (8E10G6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61697
This item is not returnable.
View return policy
Description
CCR1 Monoclonal specifically detects CCR1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| CCR1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| C-C chemokine receptor type 1, CC-CKR-1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R | |
| Mouse | |
| 41.2 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 8E10G6 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| CCR1 | |
| Purified recombinant fragment of human CD191 (AA: extra mix) expressed in E. Coli. | |
| Protein G purified | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 1230 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction