missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554994
This item is not returnable.
View return policy
Description
Immunogen sequence: DYSDRTISDE DESDEDMFMK FVSEDLHRCA LLTADSFGDP FFPRTTQILL EYQLGRWVPR LREPRDLYGV SSSGPLSPTR WPYHCEVIDE Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 90%.
Metallocarboxypeptidase that mediates both deglutamylation and deaspartylation of target proteins. Catalyzes the deglutamylation of polyglutamate side chains generated by post-translational polyglutamylation in proteins such as tubulins. Also removes gene-encoded polyglutamates or polyaspartates from the carboxy-terminus of target proteins such as MYLK. Does not show detyrosinase or deglycylase activities from the carboxy-terminus of tubulin. [UniProt].
Specifications
| CCP3 | |
| Polyclonal | |
| Unconjugated | |
| Agbl3 | |
| 2900053G10Rik; 4930431N21Rik; 6530406M24Rik; Agbl3; ATP/GTP binding protein like 3; ATP/GTP binding protein-like 3; ATP/GTP-binding protein-like 3; CBCP3; CCP3; Cytosolic carboxypeptidase 3; RGD1561995 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 340351 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NEM8 | |
| Agbl3 | |
| Recombinant protein corresponding to Human CCP3. Recombinant protein control fragment (Product #RP-92758). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction