missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCP2 Monoclonal Antibody (1F3C12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA550280
This item is not returnable.
View return policy
Description
CCP2 Monoclonal Antibody for Western Blot, ELISA
Metallocarboxypeptidase that mediates deglutamylation of target proteins. Catalyzes the deglutamylation of polyglutamate side chains generated by post-translational polyglutamylation in proteins such as tubulins. Also removes gene-encoded polyglutamates from the carboxy-terminus of target proteins such as MYLK. Does not show detyrosinase or deglycylase activities from the carboxy-terminus of tubulin.
Specifications
| CCP2 | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Q5U5Z8 | |
| AGBL2 | |
| Recombinant Human Cytosolic carboxypeptidase 2 protein (10-270AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA, Western Blot | |
| 1F3C12 | |
| Unconjugated | |
| AGBL2 | |
| 4930524K04; A430081C19Rik; Agbl2; ATP/GTP binding protein like 2; ATP/GTP binding protein-like 2; ATP/GTP-binding protein-like 2; carboxypeptidase 2, cytosolic; CBCP2; CCP2; cytoplasmic carboxypeptidase 2; cytosolic carboxypeptidase 2; testis tissue sperm-binding protein Li 96mP; testis-expressed protein 25; zebrafish testis-expressed 25; zgc:165648; zte25 | |
| Mouse | |
| Protein G | |
| RUO | |
| 79841 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction