missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCNYL3 Polyclonal Antibody

Description
CCNYL3 Polyclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | CCNYL3 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | CCNYL3 |
| Gene Accession No. | P0C7X3 |
| Gene Alias | CCNYL3; LOC102724928; putative cyclin-Y-like protein 3 |
| Gene Symbols | CCNYL3 |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?