Learn More
CCNY Mouse anti-Human, Unconjugated, Clone: 2C9E3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66865-1-IG-20UL
This item is not returnable.
View return policy
Description
The CCNY antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CCNY. This antibody recognizes human, rat, mouse antigen. The CCNY antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| CCNY | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunofluorescence, ELISA | |
| 2C9E3 | |
| Unconjugated | |
| 20 μL | |
| 219771 | |
| IgG1 |