missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCNL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13138-1-AP-150UL
This item is not returnable.
View return policy
Description
FUS/TLS was originally described as fused in Sarcoma is actually related to a number of different cellular processes and correlated to several diseases. FUS binds DNA as a single or double strand, and acts as a site for transcription factors to recognize. Since FUS is well-documented to be involved in liposarcoma, lipoma and leukaemia, it is believed that when its normal housekeeping functions related to transcription are disrupted, normal cells become cancerous. Interestingly, FUS is also associated with the fatal neuron disorder amyotrophic lateral sclerosis; interruption of its normal RNA-binding functions leads to incorrect cellular localization and leads to pathologies of the neuron. Thus, FUS regulation of cellular functions is a critical component of maintece of healthy cells.Specifications
| CCNL1 | |
| Polyclonal | |
| Unconjugated | |
| CCNL1 | |
| ania 6a, BM 001, CCNL1, Cyclin L, cyclin L1, PRO1073, UNQ530/PRO1073 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114121, 56706, 57018 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q52KE7, Q9R1Q2, Q9UK58 | |
| Ccnl1 | |
| CCNL1 Fusion Protein Ag3731 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction