missing translation for 'onlineSavingsMsg'
Learn More

CCNK Antibody (S1), Novus Biologicals™

Product Code. 18350759 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18350759 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18350759 Supplier Novus Biologicals Supplier No. H00008812M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCNK Monoclonal antibody specifically detects CCNK in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCNK
Applications Western Blot, ELISA
Classification Monoclonal
Clone S1
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH15935
Gene Alias CPR4MGC9113, cyclin K, cyclin-K
Host Species Mouse
Immunogen CCNK (AAH15935, 1 a.a. ~ 354 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MKENKENSSPSVTSANLDHTKPCWYWDKKDLAHTPSQLEGLDPATEARYRREGARFIFDVGTRLGLHYDTLATGIIYFHRFYMFHSFKQFPRYVTGACCLFLAGKVEETPKKCKDIIKTARSLLNDVQFGQFGDDPKEEVMVLERILLQTIKFDLQVEHPYQFLLKYAKQLKGDKNKIQKLVQMAWTFVNDSLCTTLSLQWEPEIIAVAVMYLAGRLCKFEIQEWTSKPMYRRWWEQFVQDVPVDVLEDICHQILDLYSQGKQQMPHHTPHQLQQPPSLQPTPQVPQVQQSQPSQSSEPSQPQQKDPLILLQGWACRQPATHLLPSPLEDSLLCPRPFPHPACLQLGGWGGQPG
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 8812
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.