missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCNK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-84391-25ul
This item is not returnable.
View return policy
Description
CCNK Polyclonal antibody specifically detects CCNK in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| CCNK | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL | |
| CPR4MGC9113, cyclin K, cyclin-K | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:DHTKPCWYWDKKDLAHTPSQLEGLDPATEARYRREGARFIFDVGTRLGLHYDTLATGIIYFHRFYMFHSFKQFPRYVTGACCLFLAGKVEETPKKCKDIIKTARSLLNDVQFGQFGDDPKEEVMVLERILLQTIKFDLQVEHPYQFLL | |
| 25 μL | |
| Cell Cycle and Replication | |
| 8812 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunocytochemistry | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction