missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CCND1-FGF19/CEN11p FISH Probe

Product Code. 16013295
Click to view available options
Quantity:
100 μL
200 μL
Unit Size:
100µL
200µL
This item is not returnable. View return policy

Product Code. 16013295

Brand: Abnova™ FG0185.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.

Specifications

Content And Storage Store at 4°C in the dark.
Description DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Format Tube
For Use With (Application) FISH-Ce
Gene ID (Entrez) 595/9965
Includes DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Quantity 100 μL
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.