missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCNC Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26464-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the cyclin family of proteins. The encoded protein interacts with cyclin-dependent kinase 8 and induces the phophorylation of the carboxy-terminal domain of the large subunit of RNA polymerase II. The level of mRNAs for this gene peaks in the G1 phase of the cell cycle. Two transcript variants encoding different isoforms have been found for this gene.Specifications
| CCNC | |
| Polyclonal | |
| Unconjugated | |
| CCNC | |
| CCNC, CycC, cyclin C, hSRB11, SRB11 homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51813, 892 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P24863, Q62447 | |
| Ccnc | |
| CCNC Fusion Protein Ag23955 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction