missing translation for 'onlineSavingsMsg'
Learn More

CCNB3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£362.00

Specifications

Antigen CCNB3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18259785
View Documents
Novus Biologicals
NBP1-80384
100 μL
£362.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CCNB3 Polyclonal specifically detects CCNB3 in Human samples. It is validated for Western Blot.
Specifications

Specifications

CCNB3
Polyclonal
Rabbit
Cell Cycle and Replication
NP_149020
85417
Synthetic peptide directed towards the C terminal of human CCNB3. Peptide sequence ISELHPLVRQLNKLLTFSSYDSLKAVYYKYSHPVFFEVAKIPALDMLKLE.
Primary
Western Blot
Unconjugated
RUO
Human
CYCB3, cyclin B3, G2/mitotic-specific cyclin-B3
CCNB3
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.