missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCNB1IP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA526407
This item is not returnable.
Se returpolicy
Beskrivning
CCNB1IP1 Polyclonal Antibody for Western Blot, IHC (P), Flow
HEI10 is a member of the E3 ubiquitin ligase family and functions in progression of the cell cycle through G(2)/M.
Specifikationer
| CCNB1IP1 | |
| Polyclonal | |
| Unconjugated | |
| CCNB1IP1 | |
| C14orf18; CCNB1IP1; cyclin B1 interacting protein 1; cyclin B1 interacting protein 1, E3 ubiquitin protein ligase; cyclin-B1-interacting protein 1; E3 ubiquitin-protein ligase CCNB1IP1; Gm288; HEI10; Human enhancer of invasion 10; Mei4; RING-type E3 ubiquitin transferase CCNB1IP1; Unknown (protein for MGC:134598) | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 57820 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9NPC3 | |
| CCNB1IP1 | |
| KLH conjugated synthetic peptide between 199-228 amino acids from the C-terminal region of human CCNB1IP1. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering