missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554173
This item is not returnable.
View return policy
Description
Immunogen sequence: LHSDDSSTKV DIKETYEVEA STFCFPESVD VGGASPHSKT ISESELSASA TELLQDYMLT LRTKLSSQEI QQFAALLHEY RNGA Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.
This gene encodes a scaffold protein that functions in the stress-activated p38 Mitogen-activated protein kinase (MAPK) signaling cascade. The protein interacts with SMAD specific E3 ubiquitin protein ligase 1 (also known as SMURF1) via a phosphotyrosine binding domain to promote RhoA degradation. The protein is required for normal cytoskeletal structure, cell-cell interactions, and lumen formation in endothelial cells. Mutations in this gene result in cerebral cavernous malformations. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| CCM2 | |
| Polyclonal | |
| Unconjugated | |
| CCM2 | |
| BC029157; C7orf22; Ccm2; CCM2 scaffolding protein; cerebral cavernous malformation 2; cerebral cavernous malformation 2 homolog; cerebral cavernous malformations 2 protein; Cerebral cavernous malformations protein 2 homolog; malcavernin; MGC4067; MGC4607; MGC74868; Oncostatin; OncostatinM; Osm; osmosensing scaffold for MEKK3; PP10187 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 83605 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BSQ5 | |
| CCM2 | |
| Recombinant protein corresponding to Human CCM2. Recombinant protein control fragment (Product #RP-92737). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction