missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCM2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-53812
This item is not returnable.
View return policy
Description
CCM2 Polyclonal specifically detects CCM2 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| CCM2 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| C7orf22OSM, cerebral cavernous malformation 2, Cerebral cavernous malformations 2 protein, chromosome 7 open reading frame 22, malcavernin, MGC4067, MGC4607, MGC74868 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize isoform 1 (NP_001025006.1), isoform 2 (NP_113631.1) and isoform 4 (NP_001161407.1). | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.3-2 ug/ml, Flow Cytometry 10 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/ml, Peptide ELISA Detection limit 1:128000 | |
| NP_001025006.1 | |
| CCM2 | |
| Peptide with sequence C-KGEKSRDKKAHEK, from the internal region of the protein sequence according to NP_001025006.1; NP_113631.1; NP_001161407.1; NP_001350387.1. | |
| 0.1 mg | |
| Signal Transduction | |
| 83605 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction