missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCM2 Antibody (OTI1E9), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72235PCP
This item is not returnable.
View return policy
Description
CCM2 Monoclonal antibody specifically detects CCM2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| CCM2 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI1E9 | |
| Western Blot, Immunohistochemistry | |
| C7orf22OSM, cerebral cavernous malformation 2, Cerebral cavernous malformations 2 protein, chromosome 7 open reading frame 22, malcavernin, MGC4067, MGC4607, MGC74868 | |
| Full length human recombinant protein of human CCM2(NP_113631) produced in HEK293T cell. | |
| 0.1 mL | |
| Signal Transduction | |
| 83605 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction