missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL8/MCP-2 Mouse anti-Human, Clone: CCL8/3686, Novus Biologicals™
Mouse Monoclonal Antibody
£216.00 - £415.00
Specifications
| Antigen | CCL8/MCP-2 |
|---|---|
| Clone | CCL8/3686 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18321883
|
Novus Biologicals
NBP3-13771-20UG |
20 μg |
£216.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18380445
|
Novus Biologicals
NBP3-13771-100UG |
100 μg |
£415.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCL8/MCP-2 Monoclonal antibody specifically detects CCL8/MCP-2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| CCL8/MCP-2 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| C-C motif chemokine 8, chemokine (C-C motif) ligand 8, HC14, MCP2, MCP-2, member 8 (monocyte chemotactic protein 2), monocyte chemoattractant protein 2, SCYA10, small-inducible cytokine A8 | |
| Human recombinant CCL8/MCP-2 protein fragment (around aa1-99) (exact sequence is proprietary) (Uniprot: P80075) | |
| Primary | |
| Store at 4°C. |
| CCL8/3686 | |
| Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P80075 | |
| 6355 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title