Learn More
PeproTech CCL6 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody
Brand: PeproTech 500-P112BT-25UG
Description
AA Sequence of recombinant protein: GLIQEIEKED RRYNPPIIHQ GFQDTSSDCC FSYATQIPCK RFIYYFPTSG GCIKPGIIFI SRRGTQVCAD PSDRRVQRCL STLKQGPRSG NKVIA. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine C10 (CCL6). Anti-Murine C10 (CCL6)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Murine C10 (CCL6) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Murine C10 (CCL6) (500-P112) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine C10 (CCL6). Western Blot: To detect mC10 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant mC-10 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P112BT-1MG will be provided as 2 x 500 μg
Chemokine(C-C motif) ligand 6(CCL6) is a small cytokine belonging to the CC chemokine family that has only been identified in rodents. In mice, CCL6 is expressed in cells from neutrophil and macrophage lineages, and can be greatly induced under conditions suitable for myeloid cell differentiation. It is highly expressed in bone marrow cultures that have been stimulated with the cytokine GM-CSF. Some low levels of gene expression also occur in certain cell lines of myeloid origin(e.g. the immature myeloid cell lines DA3 and 32D cl3, and the macrophage cell line P388D) that can also be greatly induced in culture with GM-CSF. However, in activated T cell lines, expression of CCL6 is greatly reduced. CCL6 can also be induced in the mouse lung by the cytokine interleukin 13. Mouse CCL6 is located on chromosome 11. The cell surface receptor for CCL6 is believed to be the chemokine receptor CCR1.
Specifications
| CCL6 | |
| Polyclonal | |
| Biotin | |
| Ccl6 | |
| c10; CC chemokine C10; C-C motif chemokine 6; Ccl6; CCL6(22-95); CCL6(23-95); chemokine (C-C motif) ligand 6; chemokine ligand 6; Mrp-1; Protein C10; Scay6; Scya6; small inducible cytokine A6; small-inducible cytokine A6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20305 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P27784 | |
| Ccl6 | |
| E.coli-derived Recombinant Murine C10 (CCL6). | |
| 25 μg | |
| Primary | |
| Mouse | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.