Learn More
PeproTech CCL5 (RANTES) Monoclonal Antibody, PeproTech®

Description
AA Sequence of recombinant protein: SPYSSDTTPC CFAYIARPLP RAHIKEYFYT SGKCSNPAVV FVTRKNRQVC ANPEKKWVRE YINSLEMS. Preparation: Produced in BALB/c mice immunized with highly pure Recombinant Human RANTES (CCL5). Anti-Human RANTES (CCL5)-specific antibody was purified from ascites fluid by Protein A affinity chromatography. Sandwich ELISA: Assuming 100 μL/well, a concentration of 2.0-4.0 μg/mL of this antibody will detect at least 500 pg/mL of Recombinant Human RANTES (CCL5) when used with PeproTech Biotinylated Antigen Affinity Purified Anti-Human RANTES (CCL5) (500-P36BT) as the detection antibody at a concentration of approximately 0.5-1.0 μg/mL. Western Blot: To detect Human RANTES (CCL5) by Western Blot analysis this antibody can be used at a concentration of 0.5-1.0 μg/mL. When used in conjunction with compatible secondary reagents the detection limit for Recombinant Human RANTES (CCL5) is 0.25-2.0 ng/lane, under reducing conditions and 0.25-0.5 ng/lane, under non-reducing conditions.
Specifications
Specifications
| Antigen | CCL5 (RANTES) |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | CCL5 |
| Gene Accession No. | P13501 |
| Gene Alias | beta-chemokine RANTES; CC chemokine; C-C motif chemokine 5; C-C motif chemokine ligand 5; CCL5; chemokine (C-C motif) ligand 5; D17S136E; EoCP; eosinophil chemotactic cytokine; MGC17164; MuRantes; OTTHUMP00000197106; RANTES; RANTES protein; RANTES(3-68); RANTES(4-68); regulated upon activation normal T-cell expressed and secreted; regulated upon activation, normally T-expressed, and presumably secreted; Scya5; SIS δ; SISd; SIS-delta; SISδ; small inducible cytokine A5; small inducible cytokine subfamily A (Cys-Cys), member 5; small-inducible cytokine A5; T cell-specific protein P228; T-cell specific protein p288; T-cell-specific protein RANTES; TCP228; Unknown (protein for MGC:127014) |
| Gene Symbols | CCL5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.