missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CCL5/RANTES Antibody Pair [HRP]

Product Code. p-200083687 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18610619

Brand: Novus Biologicals™ NBP31165215Plates

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pair [HRP]

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.78 to 50 pg/mL (example only, lot dependent)
Assay Sensitivity 0.78 pg/mLl (example only; lot dependent)
Conjugate HRP
Detection Method Sandwich ELISA
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Sandwich ELISA
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CCL5/RANTES.
Target Species Human
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 6352
Gene Symbol CCL5
Quantity 15 Plates
Research Discipline Biologically Active Proteins, Cancer, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym CCL5, chemokine (C-C motif) ligand 5, D17S136Enormally T-expressed, and presumably secreted, EoCP, Eosinophil chemotactic cytokine, RANTES/CCL5, SIS-delta, small inducible cytokine A5 (RANTES), small inducible cytokine subfamily A (Cys-Cys), member 5, Small-inducible cytokine A5, T cell-specific protein P228, T-cell specific protein p288, TCP228T-cell-specific protein RANTES
Target Human
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.