missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL5/RANTES Antibody (NI-0701) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28135JF669
This item is not returnable.
View return policy
Description
CCL5/RANTES Monoclonal antibody specifically detects CCL5/RANTES in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CCL5/RANTES | |
| Monoclonal | |
| Janelia Fluor 669 | |
| chemokine (C-C motif) ligand 5, D17S136Enormally T-expressed, and presumably secreted, EoCP, Eosinophil chemotactic cytokine, SIS-delta, small inducible cytokine A5 (RANTES), small inducible cytokine subfamily A (Cys-Cys), member 5, Small-inducible cytokine A5, T cell-specific protein P228, T-cell specific protein p288, TCP228T-cell-specific protein RANTES | |
| CCL5/ RANTES | |
| 0.1 mL | |
| Biologically Active Proteins, Cancer, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity | |
| 6352 | |
| Store at 4°C in the dark. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| NI-0701 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction